Diacethyl
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2,3-Butanedione
OR5248-100ML
OR5248-500ML
APOSOR5248-100MLEA
1670
CZK
APOSOR5248-100ML
APOSOR5248-500ML
Diacethyl
Diacethyl
Vzorec:
C₄H₆O₂ MW: 86.09 Bod varu: 88 °C Bod tání: –4…–2 °C Hustota: 0.981 g/cm³ (25 °C) Bod vzplanutí: 44476 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008756 CAS číslo: 431-03-8 EINECS: 207-069-8 UN: 2346 ADR: 3 II Merck Index: 13,02985 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...