2-Bromo-5-methoxybenzyl bromide
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-Bromo-3-(bromomethyl)anisole, 1-Bromo-2-(bromomethyl)-4-methoxybenzene
OR52426-100G
OR52426-25G
OR52426-5G
APOSOR52426-100GEA
5390
CZK
APOSOR52426-100G
APOSOR52426-25G
APOSOR52426-5G
2-Bromo-5-methoxybenzyl bromide
2-Bromo-5-methoxybenzyl bromide
Vzorec:
C₈H₈Br₂O MW: 279.96 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00078664 CAS číslo: 19614-12-1 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...