Tisk…

Benzyl(3-oxocyclobutyl)carbamate 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Aminocyclobutanone, N-CBZ protected, Benzyl (3-oxocyclobutyl)carbamate

OR52418-10G OR52418-1G OR52418-5G
APOSOR52418-10GEA 9790 CZK
APOSOR52418-10G APOSOR52418-1G APOSOR52418-5G
Benzyl(3-oxocyclobutyl)carbamate 97%
Benzyl(3-oxocyclobutyl)carbamate
Vzorec: C₁₂H₁₃NO₃
MW: 219.2400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13152267
CAS číslo: 130369-36-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR