Tisk…

(S)-Morpholine-3-carboxylic acid hydrochloride

Dodavatel: Apollo Scientific
Varování

OR52358-5G
APOSOR52358-5GEA 4270 CZK
APOSOR52358-5G
(S)-Morpholine-3-carboxylic acid hydrochloride
(S)-Morpholine-3-carboxylic acid hydrochloride
Vzorec: C₅H₁₀ClNO₃
MW: 167,5900 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06809590
CAS číslo: 1187929-04-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR