Tisk…

1-[(Ammoniooxy)methyl]-4-methoxybenzene chloride

Dodavatel: Apollo Scientific
Varování

OR52352-1G OR52352-5G
APOSOR52352-1GEA 695 CZK
APOSOR52352-1G APOSOR52352-5G
1-[(Ammoniooxy)methyl]-4-methoxybenzene chloride
1-[(Ammoniooxy)methyl]-4-methoxybenzene chloride
Vzorec: C₈H₁₂ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD01114582
CAS číslo: 876-33-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR