Tisk…

Ethyl malonyl chloride 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR5235-50G OR5235-100G
APOSOR5235-50GEA 5820 CZK
APOSOR5235-50G APOSOR5235-100G
Ethyl malonyl chloride 97%
Ethyl malonyl chloride
Vzorec: C₅H₇ClO₃
Bod varu: 79…80 °C/25mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00000736
CAS číslo: 36239-09-5
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR