2-(Aminomethyl)-5-bromopyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(5-Bromopyridin-2-yl)methylamine, (5-Bromopyridin-2-yl)methanamine
OR52328-1G
OR52328-5G
APOSOR52328-1GEA
4240
CZK
APOSOR52328-1G
APOSOR52328-5G
2-(Aminomethyl)-5-bromopyridine
2-(Aminomethyl)-5-bromopyridine
Vzorec:
C₆H₇BrN₂ MW: 187,0400 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06213858 CAS číslo: 173999-23-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...