3-(1H-Pyrazol-5-yl)aniline 96%
Dodavatel: Apollo Scientific
|
Varování
|
OR5227-10G
OR5227-5G
OR5227-1G
APOSOR5227-10GEA
22580
CZK
APOSOR5227-10G
APOSOR5227-5G
APOSOR5227-1G
3-(1H-Pyrazol-5-yl)aniline 96%
3-(1H-Pyrazol-5-yl)aniline
Vzorec:
C₉H₉N₃ Storage Temperature: Ambient |
MDL Number:
MFCD09055176 CAS číslo: 89260-46-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...