Tisk…

2,4,6-Trimethylphenylboronic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2,4,6-Trimethylbenzeneboronic acid, (2,4,6-Trimethylphenyl)metaboric acid, Dihydroxy(2,4,6-trimethylphenyl)borane

OR5226-100G OR5226-25G OR5226-5G
APOSOR5226-100GEA 6780 CZK
APOSOR5226-100G APOSOR5226-25G APOSOR5226-5G
2,4,6-Trimethylphenylboronic acid 97%
2,4,6-Trimethylphenylboronic acid
Vzorec: C₉H₁₃BO₂
MW: 164.01
Bod tání: 127…129 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00236060
CAS číslo: 5980-97-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR