Tisk…

Bis[(pinacolato)boryl]methane 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2,2'-Methylenebis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane), Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)methane

OR52256-1G OR52256-5G OR52256-25G
APOSOR52256-1GEA 744 CZK
APOSOR52256-1G APOSOR52256-5G APOSOR52256-25G
Bis[(pinacolato)boryl]methane 95%
Bis[(pinacolato)boryl]methane

Uses: A Useful Methylene Unit Usable for Suzuki-Miyaura Cross-coupling ;One-Pot Synthesis of Symmetrical and Unsymmetrical Diarylmethanes; ?-Alkylation of bis[(pinacolato)boryl]methane and its application to the asymmetric Suzuki-Miyaura cross-coupling

Vzorec: C₁₃H₂₆B₂O₄
MW: 267.97 g/mol
Bod tání: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD27977747
CAS číslo: 78782-17-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR