2-Bromothiophene-3-carbonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR5220-1G
OR5220-250MG
APOSOR5220-1GEA
13560
CZK
APOSOR5220-1G
APOSOR5220-250MG
2-Bromothiophene-3-carbonyl chloride
2-Bromothiophene-3-carbonyl chloride
Vzorec:
C₅H₂BrClOS Bod tání: 61…62 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09064979 CAS číslo: 197370-13-1 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...