Tisk…

Methyl-3-amino-5-chlorobenzoate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 3-amino-5-chlorobenzoate

OR52184-1G OR52184-5G
APOSOR52184-1GEA 2530 CZK
APOSOR52184-1G APOSOR52184-5G
Methyl-3-amino-5-chlorobenzoate 98%
Methyl-3-amino-5-chlorobenzoate
Vzorec: C₈H₈ClNO₂
MW: 185,6076 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11053764
CAS číslo: 21961-31-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR