Tisk…

1,3-Oxazole-5-carbaldehyde

Dodavatel: Apollo Scientific
Varování

Synonyms: Oxazole-5-carbaldehyde

OR52173-1G OR52173-250MG
APOSOR52173-1GEA 10820 CZK
APOSOR52173-1G APOSOR52173-250MG
1,3-Oxazole-5-carbaldehyde
1,3-Oxazole-5-carbaldehyde
Vzorec: C₄H₃NO₂
Storage Temperature: Freezer
MDL Number: MFCD02179513
CAS číslo: 118994-86-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR