4-Dodecylbenzenesulphonic acid
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
P-Dodecylbenzenesulphonic acid
OR52171-100G
OR52171-5G
OR52171-25G
APOSOR52171-100GEA
1990
CZK
APOSOR52171-100G
APOSOR52171-5G
APOSOR52171-25G
4-Dodecylbenzenesulphonic acid
4-Dodecylbenzenesulphonic acid
Vzorec:
C₁₈H₃₀O₃S MW: 326,49 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00066677 CAS číslo: 121-65-3 UN: 2586 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...