Glycine methyl ester hydrochloride (H-Gly-OMe.HCl) 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl glycinate hydrochloride, H-Gly-OMe.HCl, Glycine Methyl Ester HCl
OR52169-100G
OR52169-1KG
APOSOR52169-100GEA
622
CZK
APOSOR52169-100G
APOSOR52169-1KG
Glycine methyl ester hydrochloride (H-Gly-OMe.HCl) 95%
Glycine methyl ester hydrochloride (H-Gly-OMe.HCl)
Vzorec:
C₃H₈ClNO₂ MW: 125,56 g/mol Bod tání: 175 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00012870 CAS číslo: 5680-79-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...