Tisk…

(-)-Corey lactone

Dodavatel: Apollo Scientific
Varování

Synonyms: (-)-6β-Hydroxymethyl-7α-hydroxy-cis-2-oxabicyclo[3.3.0]octan-3-one, (3aR,4S,5R,6aS)-5-Hydroxy-4-(hydroxymethyl)hexahydro-2H-cyclopenta[b]furan-2-one, (3aR,4S,5R,6aS)-(-)-Hexahydro-5-hydroxy-4-(hydroxymethyl)-2H-cyclopenta[b]furan-2-one, (-)-Corey lactone diol

OR52144-5G
APOSOR52144-5GEA 3410 CZK
APOSOR52144-5G
(-)-Corey lactone
(-)-Corey lactone
Vzorec: C₈H₁₂O₄
MW: 172,18 g/mol
Bod tání: 115…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00075234
CAS číslo: 32233-40-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR