Tisk…

4,4'-Bis(2-methoxystyryl)biphenyl

Dodavatel: Apollo Scientific
Varování

Synonyms: 4,4-Bis(2-methoxystyryl)biphenyl, 4,4'-Bis[2-(2-methoxyphenyl)vinyl]biphenyl

OR52141-1KG OR52141-500G OR52141-100G
APOSOR52141-1KGEA 4200 CZK
APOSOR52141-1KG APOSOR52141-500G APOSOR52141-100G
4,4'-Bis(2-methoxystyryl)biphenyl
4,4'-Bis(2-methoxystyryl)biphenyl

Optical brightener. Recommended for use in a wide variety of plastics such as PVC and PP. Also for use with artificial leathers.

Vzorec: C₃₀H₂₆O₂
Bod tání: 216…222 °C
Storage Temperature: Ambient
MDL Number: MFCD23160777
CAS číslo: 40470-68-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR