2-(3-Butynyloxy)tetrahydro-2H-pyran 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(But-3-yn-1-yloxy)tetrahydro-2H-pyran
OR52129-25G
OR52129-5G
APOSOR52129-25GEA
6390
CZK
APOSOR52129-25G
APOSOR52129-5G
2-(3-Butynyloxy)tetrahydro-2H-pyran 97%
2-(3-Butynyloxy)tetrahydro-2H-pyran
Vzorec:
C₉H₁₄O₂ MW: 154,209 g/mol Bod varu: 92…95 °C/18mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00012352 CAS číslo: 40365-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...