Bromoacetonitrile 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR52127-100G
OR52127-25G
OR52127-500G
APOSOR52127-100GEA
2070
CZK
APOSOR52127-100G
APOSOR52127-25G
APOSOR52127-500G
Bromoacetonitrile 97%
Bromoacetonitrile
Vzorec:
BrCNCH₂ Bod varu: 148…150 °C Hustota: 1,722 g/cm³ (25 °C) Bod vzplanutí: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001884 CAS číslo: 590-17-0 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...