Tisk…

3-Amino-4-iodopyridine 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-Iodo-3-pyridylamine, 4-Iodopyridin-3-amine, 4-Iodo-pyridin-3-ylamine

OR52091-5G OR52091-25G
APOSOR52091-5GEA 2610 CZK
APOSOR52091-5G APOSOR52091-25G
3-Amino-4-iodopyridine 95%
3-Amino-4-iodopyridine
Vzorec: C₅H₅IN₂
MW: 220,01 g/mol
Bod tání: 58…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04971334
CAS číslo: 105752-11-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR