Tisk…

Cyclopropanecarboxaldehyde 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Cyclopropanecarbaldehyde

OR52083-100G OR52083-25G OR52083-5G
APOSOR52083-100GEA 4240 CZK
APOSOR52083-100G APOSOR52083-25G APOSOR52083-5G
Cyclopropanecarboxaldehyde 98%
Cyclopropanecarboxaldehyde
Vzorec: C₄H₆O
MW: 70,09 g/mol
Bod varu: 98…101 °C
Hustota: 0,938 g/cm³ (25 °C)
Bod vzplanutí: 7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012261
CAS číslo: 1489-69-6
UN: 2924
ADR: 3+8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR