Tisk…

5-Bromoquinoline-2-carboxylic acid 97%

Dodavatel: Apollo Scientific
Varování

OR52061-1G OR52061-250MG
APOSOR52061-1GEA 8070 CZK
APOSOR52061-1G APOSOR52061-250MG
5-Bromoquinoline-2-carboxylic acid 97%
5-Bromoquinoline-2-carboxylic acid
Vzorec: C₁₀H₆BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD09881345
CAS číslo: 1017412-53-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR