3'-Bromo-2'-methoxyacetophenone 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR52056-1G
OR52056-250MG
OR52056-5G
APOSOR52056-1GEA
827
CZK
APOSOR52056-1G
APOSOR52056-250MG
APOSOR52056-5G
3'-Bromo-2'-methoxyacetophenone 97%
3'-Bromo-2'-methoxyacetophenone
Vzorec:
C₉H₉BrO₂ Storage Temperature: Ambient |
MDL Number:
MFCD22573042 CAS číslo: 267651-23-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...