6-(Hydroxymethyl)picolinic acid 95+%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-(Hydroxymethyl)pyridine-2-carboxylic acid
OR52053-1G
OR52053-5G
APOSOR52053-1GEA
3910
CZK
APOSOR52053-1G
APOSOR52053-5G
6-(Hydroxymethyl)picolinic acid 95+%
6-(Hydroxymethyl)picolinic acid
Vzorec:
C₁H₁NO₃ MW: 153.1354 g/mol Bod tání: 136…137 °C Storage Temperature: Ambient |
MDL Number:
MFCD09032021 CAS číslo: 1197-10-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...