3-(4-Methylpiperazin-1-yl)propylamine 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1-(3-Aminopropyl)-4-methylpiperazine
OR52045-1G
OR52045-25G
OR52045-5G
APOSOR52045-1GEA
2780
CZK
APOSOR52045-1G
APOSOR52045-25G
APOSOR52045-5G
3-(4-Methylpiperazin-1-yl)propylamine 98%
3-(4-Methylpiperazin-1-yl)propylamine
Vzorec:
C₈H₁₉N₃ MW: 157,26 g/mol Bod varu: 113…114 °C/13mmHg Hustota: 0,924 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00014616 CAS číslo: 4572-03-6 EINECS: 224-954-4 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...