Tisk…

(6-Methoxypyridin-3-yl)methylamine

Dodavatel: Apollo Scientific
Varování

Synonyms: (6-Methoxypyridin-3-yl)methanamine, 5-(Aminomethyl)-2-methoxypyridine

OR52039-1G OR52039-250MG
APOSOR52039-1GEA 1660 CZK
APOSOR52039-1G APOSOR52039-250MG
(6-Methoxypyridin-3-yl)methylamine
(6-Methoxypyridin-3-yl)methylamine
Vzorec: C₇H₁₀N₂O
MW: 138,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06212825
CAS číslo: 262295-96-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR