Tisk…

Methyl-3-bromoisonicotinate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 3-bromoisonicotinate

OR51979-100G OR51979-25G OR51979-5G
APOSOR51979-100GEA 6880 CZK
APOSOR51979-100G APOSOR51979-25G APOSOR51979-5G
Methyl-3-bromoisonicotinate 98%
Methyl-3-bromoisonicotinate
Vzorec: C₇H₆BrNO₂
MW: 216,0300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788226
CAS číslo: 59786-31-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR