Tisk…

4-(6-Amino-3-pyridyl)-1-boc-piperazine 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Boc-4-(6-amino-3-pyridyl)piperazine, tert-Butyl 4-(6-Amino-3-pyridyl)piperazine-1-carboxylate, tert-Butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate, 1-tert-Butoxycarbonyl-4-(6-amino-3-pyridyl)piperazine, 4-(6-Amino-3-pyridyl)piperazine-1-carboxylic acid tert-Butyl ester

OR51957-100G
APOSOR51957-100GEA 1820 CZK
APOSOR51957-100G
4-(6-Amino-3-pyridyl)-1-boc-piperazine 98%
4-(6-Amino-3-pyridyl)-1-boc-piperazine
Vzorec: C₁₄H₂₂N₂O₄
MW: 278,35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11594962
CAS číslo: 571188-59-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR