3-Chloro-5-hydroxybenzaldehyde 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR51926-5G
APOSOR51926-5GEA
6120
CZK
APOSOR51926-5G
3-Chloro-5-hydroxybenzaldehyde 98%
3-Chloro-5-hydroxybenzaldehyde
Vzorec:
C₇H₅ClO₂ MW: 156,5664 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08234659 CAS číslo: 1829-33-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...