Azomethine H sodium salt hydrate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Sodium 5-hydroxy-4-((2-hydroxybenzylidene)amino)-7-sulphonaphthalene-2-sulphonate xhydrate, Azomethine-H monosodium salt hydrate, 8-Hydroxy-1-(salicylideneamino)naphthalene-3,6-disulphonic acid 6-sodium salt hydrate, Sodium hydrogen 4-hydroxy-5-(salicylideneamino)naphthalene-2,7-disulphonate hydrate, Azomethine-H sodium hydrate
OR51919-100G
OR51919-25G
APOSOR51919-100GEA
10190
CZK
APOSOR51919-100G
APOSOR51919-25G
Azomethine H sodium salt hydrate
Azomethine H sodium salt hydrate
Reagent for boron determination.
Vzorec:
C₁₇H₁₂NNaO₈S₂·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00149588 CAS číslo: 206752-32-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...