3-Nitrophenylhydrazinium chloride
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Nitrophenylhydrazine hydrochloride, 1-(3-Nitrophenyl)hydrazine hydrochloride, (3-Nitrophenyl)hydrazine hydrochloride
OR5189-25G
OR5189-5G
APOSOR5189-25GEA
4570
CZK
APOSOR5189-25G
APOSOR5189-5G
3-Nitrophenylhydrazinium chloride
3-Nitrophenylhydrazinium chloride
Vzorec:
C₆H₈ClN₃O₂ MW: 189,6 g/mol Bod tání: 210 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00012939 CAS číslo: 636-95-3 EINECS: 211-270-6 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...