Tisk…

5-Isoxazolecarboxylic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Isoxazole-5-carboxylic acid

OR5183-25G OR5183-1G OR5183-5G
APOSOR5183-25GEA 6450 CZK
APOSOR5183-25G APOSOR5183-1G APOSOR5183-5G
5-Isoxazolecarboxylic acid 98%
5-Isoxazolecarboxylic acid
Vzorec: C₄H₃NO₃
MW: 113,07 g/mol
Bod tání: 141…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00156151
CAS číslo: 21169-71-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR