Tisk…

4'-Ethynylacetophenone 94%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(4-Ethynyl-phenyl)-ethanone, 1-(4-Ethynylphenyl)ethanone, 1 - (4 - Ethynyl - phenyl) - ethanone, Ethanone,1-(4-ethynylphenyl)-

OR51810-10G OR51810-1G OR51810-5G
APOSOR51810-10GEA 17430 CZK
APOSOR51810-10G APOSOR51810-1G APOSOR51810-5G
4'-Ethynylacetophenone 94%
4'-Ethynylacetophenone
Vzorec: C₁₀H₈O
Bod tání: 68…71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04974025
CAS číslo: 42472-69-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR