Tisk…

2-Methylterephthalic acid 98%

Dodavatel: Apollo Scientific
Varování

OR51802-250MG OR51802-1G OR51802-5G
APOSOR51802-250MGEA 744 CZK
APOSOR51802-250MG APOSOR51802-1G APOSOR51802-5G
2-Methylterephthalic acid 98%
2-Methylterephthalic acid
Vzorec: C₉H₈O₄
Storage Temperature: Ambient
MDL Number: MFCD09835103
CAS číslo: 5156-01-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR