Tisk…

2,4,5-Triamino-6-hydroxypyrimidine sulphate

Dodavatel: Apollo Scientific
Varování

OR51801-1G OR51801-5G OR51801-25G
APOSOR51801-1GEA 1580 CZK
APOSOR51801-1G APOSOR51801-5G APOSOR51801-25G
2,4,5-Triamino-6-hydroxypyrimidine sulphate
2,4,5-Triamino-6-hydroxypyrimidine sulphate
Vzorec: C₄H₉N₅O₅S
Storage Temperature: Ambient
MDL Number: MFCD00012785
CAS číslo: 39267-74-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR