Tisk…

4-Methylpyridin-3-ol

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Hydroxy-4-methylpyridine

OR51773-1G OR51773-5G OR51773-10G
APOSOR51773-1GEA 3150 CZK
APOSOR51773-1G APOSOR51773-5G APOSOR51773-10G
4-Methylpyridin-3-ol
4-Methylpyridin-3-ol
Vzorec: C₆H₇NO
MW: 109,1259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114227
CAS číslo: 1121-19-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR