5-Hydroxypicolinaldehyde 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR51764-250MG
OR51764-1G
APOSOR51764-250MGEA
622
CZK
APOSOR51764-250MG
APOSOR51764-1G
5-Hydroxypicolinaldehyde 98%
5-Hydroxypicolinaldehyde
Vzorec:
C6H5NO2 Storage Temperature: Ambient |
MDL Number:
MFCD10697538 CAS číslo: 31191-08-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...