Tisk…

N-Hydroxyphthalimide

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Hydroxyisoindoline-1,3-dione, 2-Hydroxy-1H-isoindole-1,3(2H)-dione, N-Hydroxypthalimide

OR51753-100G OR51753-2.5KG OR51753-500G
APOSOR51753-100GEA 622 CZK
APOSOR51753-100G APOSOR51753-2.5KG APOSOR51753-500G
N-Hydroxyphthalimide
N-Hydroxyphthalimide
Vzorec: C₈H₅NO₃
MW: 163,13 g/mol
Bod tání: 233 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00005891
CAS číslo: 524-38-9
EINECS: 208-358-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR