Tisk…

2-Chloro-4-methylbenzoic acid

Dodavatel: Apollo Scientific
Varování

OR5156-1G OR5156-25G OR5156-5G
APOSOR5156-1GEA 622 CZK
APOSOR5156-1G APOSOR5156-25G APOSOR5156-5G
2-Chloro-4-methylbenzoic acid
2-Chloro-4-methylbenzoic acid
Vzorec: C₈H₇ClO₂
MW: 170,5900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094667
CAS číslo: 7697-25-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR