(S)-Propane-1,2-diamine dihydrochloride
Dodavatel: Apollo Scientific
Synonyms:
(S)-1,2-Diaminopropane dihydrochloride, (S)-(-)-1,2-Diaminopropane dihydrochloride, S-(-)-Propylenediamine dihydrochloride, (S)-Propylenediamine dihydrochloride, (S)-1,2-Propanediamine dihydrochloride
OR51195-100G
APOSOR51195-100GEA
4270
CZK
APOSOR51195-100G
(S)-Propane-1,2-diamine dihydrochloride
(S)-Propane-1,2-diamine dihydrochloride
Vzorec:
C₃H₁₂Cl₂N₂ MW: 147,05 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00050706 CAS číslo: 19777-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...