1,3-Diiodo-5,5-dimethylhydantoin 97%
Dodavatel: Apollo Scientific
OR51176-25G
OR51176-5G
APOSOR51176-25GEA
10350
CZK
APOSOR51176-25G
APOSOR51176-5G
1,3-Diiodo-5,5-dimethylhydantoin 97%
1,3-Diiodo-5,5-dimethylhydantoin
Vzorec:
C₅H₆I₂N₂O₂ Bod tání: 198 °C Storage Temperature: Freezer |
MDL Number:
MFCD00020867 CAS číslo: 2232-12-4 UN: 3085 ADR: 5.1+8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...