1H-Indazole-3-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Indazole-3-carboxylic acid
OR5104-100G
OR5104-500G
APOSOR5104-100GEA
2440
CZK
APOSOR5104-100G
APOSOR5104-500G
1H-Indazole-3-carboxylic acid
1H-Indazole-3-carboxylic acid
Vzorec:
C₈H₆N₂O₂ MW: 162.15 g/mol Bod tání: 266…270 °C Storage Temperature: Ambient |
MDL Number:
MFCD00211066 CAS číslo: 4498-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...