Tisk…

2-Methoxy-9,9-dimethyl-9H-fluorene 98%

Dodavatel: Apollo Scientific

OR51025-5G
APOSOR51025-5GEA 5560 CZK
APOSOR51025-5G
2-Methoxy-9,9-dimethyl-9H-fluorene 98%
2-Methoxy-9,9-dimethyl-9H-fluorene
Vzorec: C₁₆H₁₆O
MW: 224,30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32263492
CAS číslo: 1514864-84-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR