Tisk…

N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine 99%

Dodavatel: Apollo Scientific

Synonyms: N,N′-Di(1-naphthyl)-N,N′-diphenyl-(1,1′-biphenyl)-4,4′-diamine, N4,N4'-Di(naphthalen-1-yl)-N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine

OR51012-1G OR51012-25G OR51012-5G
APOSOR51012-1GEA 622 CZK
APOSOR51012-1G APOSOR51012-25G APOSOR51012-5G
N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine 99%
N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine
Vzorec: C₄₄H₃₂N₂
MW: 588,75 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03093246
CAS číslo: 123847-85-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR