Tisk…

4-Bromo-9,9'-spirobi[9H-fluorene] 98%

Dodavatel: Apollo Scientific

Synonyms: 4-Bromo-9,9'-spirobi[fluorene]

OR51009-1G OR51009-25G OR51009-5G
APOSOR51009-1GEA 622 CZK
APOSOR51009-1G APOSOR51009-25G APOSOR51009-5G
4-Bromo-9,9'-spirobi[9H-fluorene] 98%
4-Bromo-9,9'-spirobi[9H-fluorene]
Vzorec: C₂₅H₁₅Br
MW: 395,30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16660920
CAS číslo: 1161009-88-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR