4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzofuran 99%
Dodavatel: Apollo Scientific
Synonyms:
2-(Dibenzo[b,d]furan-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, Dibenzofuran-4-boronic acid Pinacol ester, 2-(Dibenzofuran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
OR51008-1G
OR51008-25G
OR51008-5G
APOSOR51008-1GEA
744
CZK
APOSOR51008-1G
APOSOR51008-25G
APOSOR51008-5G
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzofuran 99%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzofuran
Vzorec:
C₁₈H₁₉BO₃ Storage Temperature: Ambient |
MDL Number:
MFCD18910632 CAS číslo: 912824-85-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...