2-Phenyl-9H-carbazole 99%
Dodavatel: Apollo Scientific
OR51005-1G
OR51005-25G
OR51005-5G
APOSOR51005-1GEA
622
CZK
APOSOR51005-1G
APOSOR51005-25G
APOSOR51005-5G
2-Phenyl-9H-carbazole 99%
2-Phenyl-9H-carbazole
Vzorec:
C₁₈H₁₃N MW: 243,31 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD26402145 CAS číslo: 88590-00-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...