Tisk…

(2-Bromo-6-nitrophenyl)acetic acid

Dodavatel: Apollo Scientific
Varování

OR50989-1G OR50989-250MG OR50989-5G
APOSOR50989-1GEA 5390 CZK
APOSOR50989-1G APOSOR50989-250MG APOSOR50989-5G
(2-Bromo-6-nitrophenyl)acetic acid
(2-Bromo-6-nitrophenyl)acetic acid
Vzorec: C₈H₆BrNO₄
Storage Temperature: Ambient
MDL Number: MFCD10566470
CAS číslo: 37777-74-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR