Ethyl-2-thiouracil-5-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Carbethoxy-2-thiouracil
OR50947-25G
OR50947-5G
APOSOR50947-25GEA
2690
CZK
APOSOR50947-25G
APOSOR50947-5G
Ethyl-2-thiouracil-5-carboxylate
Ethyl-2-thiouracil-5-carboxylate
Vzorec:
C₇H₈N₂O₃S MW: 200,22 g/mol Bod tání: 249…255 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006074 CAS číslo: 38026-46-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...