Tisk…

6-Hydroxynicotinic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Oxo-1,6-dihydropyridine-3-carboxylic acid, 6-Hydroxy-3-pyridinecarboxylic acid, 6-Hydroxyniacin, 6-Hydroxypyridine-3-carboxylic acid

OR5072-100G OR5072-25G OR5072-500G
APOSOR5072-100GEA 2700 CZK
APOSOR5072-100G APOSOR5072-25G APOSOR5072-500G
6-Hydroxynicotinic acid 98%
6-Hydroxynicotinic acid
Vzorec: C₆H₅NO₃
MW: 139,11 g/mol
Bod tání: 314…316 °C
Storage Temperature: Ambient
MDL Number: MFCD00006277
CAS číslo: 5006-66-6
EINECS: 225-682-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR